Phosphoryl trichloride ≥99%, ReagentPlus®

Danger

201170-1KG 201170-250G 201170-5G
220042-516EA 184.62 USD
220042-516 220042-517 220042-518
Phosphoryl trichloride ≥99%, ReagentPlus®
Phosphoryl trichloride
Formula: POCl₃
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: 1 °C
Density: 1.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011443
CAS Number: 10025-87-3
EINECS: 233-046-7
UN: 1810
ADR: 8,I
Merck Index: 12,07506

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR