Osmium tetraoxide, ReagentPlus®

Danger

201030-1G 201030-250MG 201030-100MG 201030-500MG
220042-430EA 252.31 USD
220042-430 220042-431 220042-429 220042-432
Osmium tetraoxide, ReagentPlus®
Osmium tetraoxide
Formula: O₄Os
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: 40 °C
Density: 1.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011150
CAS Number: 20816-12-0
EINECS: 244-058-7
UN: 2471
ADR: 6.1,I
Merck Index: 13,06962

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR