Lithium aluminum hydride ≥95%, Reagent Grade

Danger

Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate

199877-1KG 199877-25G 199877-100G 199877-10G
220042-183EA 720 USD
220042-183 220042-184 220042-181 220042-182
Lithium aluminum hydride ≥95%, Reagent Grade
Lithium aluminum hydride
Formula: H₄AlLi
MW: 37.95 g/mol
Melting Pt: 125 °C
Density: 0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1410
ADR: 4.3,I
Merck Index: 13,00348

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR