Picric acid ≥98% stabilized, Reagent Grade

Danger

Synonyms: 2,4,6-Trinitrophenol, TNP

197378-100G 197378-500G
220042-088EA 60.31 USD
220042-088 220042-089
Picric acid ≥98% stabilized, Reagent Grade
Picric acid
Formula: C₆H₃N₃O₇
MW: 229.11 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 80.35 °C
Density: 1.763 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00007102
CAS Number: 88-89-1
EINECS: 201-865-9
UN: 154
ADR: 1,
Merck Index: 12,07562

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR