1,3-Dimethylimidazolium methyl sulfate for catalysis

Danger

Synonyms: 1,3-Dimethyl-1H-imidazol-3-ium methyl sulphate

19409-25G 19409-5G
220041-854EA 107.54 USD
220041-854 220041-855
1,3-Dimethylimidazolium methyl sulfate for catalysis
1,3-Dimethylimidazolium methyl sulphate
Formula: C₆H₁₂N₂O₄S
MW: 208.24 g/mol
MDL Number: MFCD03453276
CAS Number: 97345-90-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR