1,4-Diiodobenzene ≥99%

193526-10G 193526-50G
220041-803EA 73.23 USD
220041-803 220041-804
1,4-Diiodobenzene ≥99%
1,4-Diiodobenzene
Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 126…127 °C
Density: 2.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001054
CAS Number: 624-38-4
EINECS: 210-842-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR