Tetrabutylammonium bromide, ReagentPlus®

193119-500G 193119-5KG 193119-25G 193119-100G
220041-099EA 137.69 USD
220041-099 220041-100 220041-098 220041-097
Tetrabutylammonium bromide, ReagentPlus®
Tetrabutylammonium bromide
Formula: C₁₆H₃₆BrN
MW: 322.37 g/mol
Melting Pt: 100…103 °C
Density: 1.148 g/cm³ (20 °C)
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR