tert-Butylchlorodimethylsilane, Reagent Grade

Danger

Synonyms: tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride

190500-5G 190500-25G 190500-100G 190500-1KG
220040-231EA 60.2 USD
220040-231 220039-563 220039-560 220039-562
tert-Butylchlorodimethylsilane, Reagent Grade
tert-Butylchlorodimethylsilane
Formula: C₆H₁₅ClSi
MW: 150.72 g/mol
Boiling Pt: 125 °C (1013 hPa)
Melting Pt: 87…90 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 22 °C
MDL Number: MFCD00000501
CAS Number: 18162-48-6
EINECS: 242-042-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR