Benzylamine ≥99%, ReagentPlus®

Danger

Synonyms: α-Aminotoluene

185701-500G 185701-5G 185701-100G
220041-039EA 81.31 USD
220041-039 220041-040 220041-038
Benzylamine ≥99%, ReagentPlus®
Benzylamine
Formula: (C₆H₅)CH₂NH₂
MW: 107.16 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 10 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00008106
CAS Number: 100-46-9
EINECS: 202-854-1
UN: 2735
ADR: 8,II
Merck Index: 13,01127

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR