Barbituric acid, ReagentPlus®

185698-100G 185698-25G 185698-500G
220041-035EA 55.54 USD
220041-035 220041-036 220041-037
Barbituric acid, ReagentPlus®
Barbituric acid
Formula: C₄H₄N₂O₃
Boiling Pt: 441 °C (1013 hPa)
Melting Pt: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00006666
CAS Number: 67-52-7
EINECS: 200-658-0
Merck Index: 13,00966

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR