Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.0%, ReagentPlus®

Danger

Synonyms: Ethylene glycol monomethyl ether

185469-500ML 185469-4L
220040-285EA 19.82 USD
220040-285 220040-284
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.0%, ReagentPlus®
Ethylene glycol monomethyl ether (2-methoxyethanol)
Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 124.6 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.964 g/cm³ (20 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR