Diisopropyl ether ≥99% stabilized, ReagentPlus®

Danger

Synonyms: Isopropyl ether

185302-4L 185302-1L
220041-019EA 80.31 USD
220041-019 220041-017
Diisopropyl ether ≥99% stabilized, ReagentPlus®
Diisopropyl ether
Formula: (CH₃)₂CHOCH(CH₃)₂
MW: 102.18 g/mol
Boiling Pt: 68.5 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.726 g/cm³ (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008880
CAS Number: 108-20-3
EINECS: 203-560-6
UN: 1159
ADR: 3,II
Merck Index: 13,05232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR