Lead tetraacetate ≥95%, Reagent Grade

Danger

Synonyms: Lead(IV) tetraacetate, Triacetyloxyplumbyl acetate, Acetic acid lead (IV) salt, Lead (IV) acetate, LTA

185191-100G 185191-500G
220040-946EA 71.77 USD
220040-946 220040-948
Lead tetraacetate ≥95%, Reagent Grade
Lead tetraacetate
Formula: (H₃CCOO)₄Pb
MW: 443.38 g/mol
Melting Pt: 175…180 °C
Density: ~ 2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008693
CAS Number: 546-67-8
EINECS: 208-908-0
UN: 1616
ADR: 6.1,III
Merck Index: 13,05442

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR