1,3-Dioxolane ≥99%, ReagentPlus®

Danger

184497-500ML 184497-4L 184497-1L
220040-769EA 34.15 USD
220040-769 220040-768 220040-767
1,3-Dioxolane ≥99%, ReagentPlus®
1,3-Dioxolane
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 75 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.066 g/cm³ (15 °C)
MDL Number: MFCD00003207
CAS Number: 646-06-0
EINECS: 211-463-5
UN: 1166
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR