1-Butyl-3-methylimidazoliumhexafluorophosphate for catalysis

Synonyms: 1-Butyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V), 1-(But-1-yl)-3-methyl-1H-imidazol-3-ium hexafluorophosphate

18122-50G-F 18122-5G-F
220040-482EA 1380 USD
220040-482 220040-483
1-Butyl-3-methylimidazoliumhexafluorophosphate for catalysis
1-Butyl-3-methylimidazoliumhexafluorophosphate
Formula: C₈H₁₅F₆N₂P
MW: 284.18 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: ∼6 °C
Density: 1.38 g/cm³ (20 °C)
Flash Pt: >110 °C
MDL Number: MFCD03093295
CAS Number: 174501-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR