Stearic acid ≥95%, Reagent Grade

Synonyms: Octadecanoic acid, Octadecylic acid

175366-1KG 175366-100G 175366-5KG
220039-746EA 55.38 USD
220039-746 220039-745 220039-747
Stearic acid ≥95%, Reagent Grade
Stearic acid
Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Boiling Pt: 361 °C (1013 hPa)
Melting Pt: 67 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00002752
CAS Number: 57-11-4
EINECS: 200-313-4
Merck Index: 13,08882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR