Sodium methoxide ≥95%, Reagent Grade

Danger

Synonyms: Sodium methylate, Sodium methanolate

164992-2KG 164992-100G 164992-5G 164992-500G
220038-660EA 136.15 USD
220038-660 220038-659 220038-662 220038-661
Sodium methoxide ≥95%, Reagent Grade
Sodium methoxide
Formula: CH₃NaO
MW: 54.02 g/mol
Melting Pt: 127 °C
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00012179
CAS Number: 124-41-4
EINECS: 204-699-5
UN: 1431
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR