Trifluoromethanesulfonic acid ≥98%, Reagent Grade

Synonyms: Trifluoromethanesulphonic acid

158534-100G 158534-10G 158534-50G
220038-407EA 133.08 USD
220038-407 220038-408 220038-409
Trifluoromethanesulfonic acid ≥98%, Reagent Grade
Trifluoromethanesulphonic acid
Formula: CHF₃O₃S
MW: 150.08 g/mol
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: 35 °C
Density: 1.69 g/cm³ (20 °C)
MDL Number: MFCD00007514
CAS Number: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR