Phosphorus(V) chloride ≥95%, Reagent Grade

Danger

157775-5G 157775-1KG 157775-100G
220038-186EA 72 USD
220038-186 220038-185 220038-846
Phosphorus(V) chloride ≥95%, Reagent Grade
Phosphorus(V) chloride
Formula: Cl₅P
Melting Pt: ∼148 °C
Density: 3.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011439
CAS Number: 10026-13-8
EINECS: 233-060-3
UN: 1806
ADR: 8,II
Merck Index: 12,07508

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR