Potassium-tert-butoxide ≥95%, Reagent Grade

Danger

Synonyms: Potassium-2-methylpropan-2-olate, Potassium tert-butylate, Potassium tert-butanolate, Potassium t-butoxide

156671-5G 156671-100G 156671-500G
220038-404EA 21.25 USD
220038-404 220038-402 220038-403
Potassium-tert-butoxide ≥95%, Reagent Grade
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
Melting Pt: 256 - 258 °C
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR