Ammonium formate ≥97%, Reagent Grade

Warning

Synonyms: Formic acid ammonium salt

156264-500G 156264-100G 156264-1KG
220038-572EA 57.05 USD
220038-572 220038-570 220038-571
Ammonium formate ≥97%, Reagent Grade
Ammonium formate
Formula: HCO₂NH₄
MW: 63.06 g/mol
Melting Pt: 119 - 121 °C
Density: 1.28 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013103
CAS Number: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR