N-Boc-N'-Fmoc-diaminoacetic acid ≥95.0% (by HPLC), Technical Grade

Danger

Synonyms: Fmoc-α-amino-DL-Gly(Boc)-OH

15513-1G-F 15513-5G-F
220038-293EA 786.15 USD
220038-293 220038-297
N-Boc-N'-Fmoc-diaminoacetic acid ≥95.0% (by HPLC), Technical Grade
N-Boc-N'-Fmoc-diaminoacetic acid
Formula: C₂₂H₂₄N₂O₆
MW: 412.442 g/mol
Melting Pt: Max. -15
Storage Temperature: Refrigerator
MDL Number: MFCD00273452
CAS Number: 176039-39-7
EINECS: 200-001-8
UN: 2209
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR