N,N-Dimethylacetamide ≥99% for spectrophotometry

Danger

154806-2L 154806-1L
220038-227EA 358.46 USD
220038-227 220038-226
N,N-Dimethylacetamide ≥99% for spectrophotometry
N,N-Dimethylacetamide
Formula: CH₃CON(CH₃)₂
Boiling Pt: 166.1 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.937 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
ADR: 6.1,III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR