1,3-Dimethylurea (symmetrical) ≥99.0% (by total nitrogen basis)

Synonyms: N,N'-Dimethylurea

15450-100G-F 15450-25G-F 15450-5G-F 15450-1KG-R
220037-260EA 154 USD
220037-260 220037-262 220037-263 220037-261
1,3-Dimethylurea (symmetrical) ≥99.0% (by total nitrogen basis)
1,3-Dimethylurea
Formula: C₃H₈N₂O
MW: 88.11 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 96 °C
Density: 1.142 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008286
CAS Number: 96-31-1
EINECS: 202-498-7

Order Now

Specification Test Results

Assay 99.0%HPLC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR