2-Methyl-2-butanol ≥99%, ReagentPlus®

Danger

Synonyms: tert-Amyl alcohol, tert-Pentyl alcohol, tert-Pentanol

152463-250ML 152463-18L 152463-1L
220037-877EA 15.63 USD
220037-877 220037-875 220037-876
2-Methyl-2-butanol ≥99%, ReagentPlus®
2-Methyl-2-butanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 102 °C (1013 hPa)
Melting Pt: -8.4 °C
Density: 0.806 g/cm³ (20 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00004478
CAS Number: 75-85-4
EINECS: 200-908-9
UN: 1105
ADR: 3,II
Merck Index: 13,07218

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR