N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane 99% (excluding TMCS), LiChropur™ for GC derivatization, Supelco®

Danger

Synonyms: BSTFA+TMCS (99:1)

15238-25ML 15238-5ML 15238-100ML
220037-869EA 281.54 USD
220037-869 220037-870 220037-868
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane 99% (excluding TMCS), LiChropur™ for GC derivatization, Supelco®
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Formula: C₈H₁₈F₃NOSi₂
MW: 257.40 g/mol
MDL Number: MFCD00008269
CAS Number: 25561-30-2
EINECS: 247-103-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR