4-Chlorophenyl isocyanate ≥98%

152277-5G 152277-100G
220037-860EA 56.11 USD
220037-860 220037-859
4-Chlorophenyl isocyanate ≥98%
4-Chlorophenyl isocyanate
Formula: C₇H₄ClNO
MW: 153.57 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 31 - 32 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002024
CAS Number: 104-12-1
EINECS: 203-176-9
UN: 2811
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR