N,O-Bis(trimethylsilyl)trifluoroacetamide ≥99.0%, LiChropur™ for GC derivatization, Sigma-Aldrich®

Danger

Synonyms: Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA

15222-10X1ML-F 15222-25ML-F 15222-1ML-F 15222-5ML-F
220037-154EA 420 USD
220037-154 220037-156 220037-155 220037-157
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥99.0%, LiChropur™ for GC derivatization, Sigma-Aldrich®
N,O-Bis(trimethylsilyl)trifluoroacetamide
Formula: C₈H₁₈F₃NOSi₂
MW: 257.4 g/mol
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: –41 °C
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00008269
CAS Number: 25561-30-2
EINECS: 247-103-9
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR