Thallium(III) trifluoroacetate, Technical Grade

Synonyms: Thallic trifluoroacetate

150533-10G 150533-50G 150533-5G
220037-661EA 110.77 USD
220037-661 220037-662 220037-663
Thallium(III) trifluoroacetate, Technical Grade
Thallium(III) trifluoroacetate
Formula: C₆F₉O₆Tl
MW: 543.43 g/mol
Boiling Pt: 160 °C (1.5 mmHg)
Melting Pt: 120.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000414
CAS Number: 23586-53-0
EINECS: 245-761-1
UN: 1707
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR