Bismarck brown R for microscopy

Synonyms: Basic Brown 4, Vesuvine, 1,3-Di((2,4-diamino-5-methylphenyl)azo)-4-methylbenzol, C.I. 21010

15000-100G 15000-25G
220036-171EA 202.34 USD
220036-171 220036-172
Bismarck brown R for microscopy
Bismarck brown R

Bismarck Brown R has been used as an adsorbate and is used in dye decoloration experiment.

Formula: C₂₁H₂₄N₈·2HCl
MW: 461.39 g/mol
MDL Number: MFCD00036386
CAS Number: 5421-66-9
EINECS: 226-541-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR