Polyoxyethylene bis(amine), M.W. 20,000, Sigma-Aldrich®

Synonyms: PEG-bis-amine, Amine-PEG-Amine, O,O′-Bis(2-aminoethyl)polyethylene glycol, PEG-diamine, Amine-PEG-amine (MW 4000), Poly(ethylene glycol) diamine, Polyoxyethylene bis(amine), Diaminopolyethylene glycol

14509-1KG-R 14509-250G-R 14509-1G-F
220037-098EA 143.31 USD
220037-098 220037-099 220037-097
Polyoxyethylene bis(amine), M.W. 20,000, Sigma-Aldrich®
Polyoxyethylene bis(amine)
Formula: H₂NCH₂CH₂(OCH₂CH₂)nNH₂
MDL Number: MFCD00132004
CAS Number: 24991-53-5
EINECS: 231-141-8
UN: 3089
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR