2,4,6-Collidine, ReagentPlus®

Danger

142387-5ML 142387-500ML 142387-100ML
220036-157EA 21.62 USD
220036-157 220036-156 220036-155
2,4,6-Collidine, ReagentPlus®
2,4,6-Collidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: –43 °C
Density: 0.9191 g/cm³ (20 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00006338
CAS Number: 108-75-8
EINECS: 203-613-3
UN: 1992
ADR: 3,III
Merck Index: 13,09789

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR