Morpholine, ReagentPlus®

Danger

134236-500ML 134236-100ML
220036-034EA 46 USD
220036-034 220036-032
Morpholine, ReagentPlus®
Morpholine
Formula: C₄H₉NO
Boiling Pt: 128.9 °C (1013 hPa)
Melting Pt: –5 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00005972
CAS Number: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8,I
Merck Index: 12,06362
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9fb9ce2adfd770be2ae8d5f63ea6220bef0f781&vid=6b2bfa885631c3f4e0681813373d468dcc171b83&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR