Cyclohexene ≥99% inhibitor-free, ReagentPlus®

Danger

125431-1L 125431-100ML
220035-064EA 28.86 USD
220035-064 220035-063
Cyclohexene ≥99% inhibitor-free, ReagentPlus®
Cyclohexene
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: -104 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: -20 °C
Storage Temperature: Ambient
MDL Number: MFCD00001539
CAS Number: 110-83-8
EINECS: 203-807-8
UN: 2256
ADR: 3,II
Merck Index: 13,02756

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR