1-Bromo-2-methylnaphthalene, Technical Grade

Warning

124109-25G 124109-100G
220034-979EA 175.54 USD
220034-979 220034-978
1-Bromo-2-methylnaphthalene, Technical Grade
1-Bromo-2-methylnaphthalene
Formula: C₁₁H₉Br
MW: 221.1 g/mol
Boiling Pt: 296 °C (1013 hPa)
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003871
CAS Number: 2586-62-1
EINECS: 219-966-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR