3,4-Dihydroxy-3-cyclobutene-1,2-dione ≥99%

Synonyms: Squaric acid, 1,2-Dihydroxy-3,4-cyclobutenedione, Quadratic acid, 1,2-Dihydroxy-1-cyclobutene-3,4-dione, Diketocyclobutenediol

123447-5G 123447-100G 123447-25G
220035-159EA 81.2 USD
220035-159 220035-157 220035-158
3,4-Dihydroxy-3-cyclobutene-1,2-dione ≥99%
3,4-Dihydroxy-3-cyclobutene-1,2-dione
Formula: C₄H₂O₄
MW: 114.06 g/mol
Melting Pt: 293 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00001334
CAS Number: 2892-51-5
EINECS: 220-761-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR