2-Amino-3-hydroxypyridin

Danger

Synonyms: 2-Aminopyridin-3-ol, 2-Amino-3-hydroxypyridine

122513-25G 122513-100G 122513-5KG
220035-027EA 135.69 USD
220035-027 220034-860 220035-028
2-Amino-3-hydroxypyridin
2-Amino-3-hydroxypyridin
Formula: C₅H₆N₂O
MW: 110.12 g/mol
Boiling Pt: 250 °C (1013 hPa)
Melting Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00006317
CAS Number: 16867-03-1
EINECS: 240-886-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR