1,1'-Carbonyldiimidazole, Reagent Grade

Danger

Synonyms: Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI

115533-100G 115533-25G 115533-25KG 115533-1KG 115533-5G 115533-10G
220034-509EA 306.92 USD
220034-509 220034-512 220034-513 220034-511 220034-514 220034-510
1,1'-Carbonyldiimidazole, Reagent Grade
1,1'-Carbonyldiimidazole
Formula: C₇H₆N₄O
MW: 162.15 g/mol
Melting Pt: 111…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00005286
CAS Number: 530-62-1
EINECS: 208-488-9
UN: 3263
ADR: 8,II
Merck Index: 14,01819

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR