Acetyl chloride ≥98%, Reagent Grade

Danger

Synonyms: Acetyl chloride 1M in Dichloromethane

114189-500G 114189-1KG 114189-25G
220034-179EA 56.15 USD
220034-179 220034-177 220034-178
Acetyl chloride ≥98%, Reagent Grade
Acetyl chloride
Formula: C₂H₃ClO
MW: 78.5 g/mol
Boiling Pt: 50.9 °C (1013 hPa)
Melting Pt: –112 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 5 °C
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 1717
ADR: 3,II
Merck Index: 13,00087

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR