1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulfonyl)imide ≥97.0% (by NMR), Sigma-Aldrich®

Synonyms: 1-ethyl-3-methylimidazolium bis((trifluoromethyl)sulphonyl)imide, [EMIIm], 1-Ethyl-3-methylimidazolium bis(trifluoromethylsulphonyl)imide

11291-1G-F 11291-5G-F
220034-402EA 217 USD
220034-402 220034-403
1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulfonyl)imide ≥97.0% (by NMR), Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide
Formula: C₈H₁₁F₆N₃O₄S₂
MW: 391.32 g/mol
MDL Number: MFCD03788927
CAS Number: 174899-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR