Benzocaine (ethyl-4-aminobenzoate) ≥98%

Warning

112909-500G 112909-100G 112909-5G
220034-400EA 52.92 USD
220034-400 220034-399 220034-401
Benzocaine (ethyl-4-aminobenzoate) ≥98%
Benzocaine (ethyl-4-aminobenzoate)
Formula: C₉H₁₁NO₂
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 89…90 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007892
CAS Number: 94-09-7
EINECS: 202-303-5
Merck Index: 13,01087

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR