N-Benzyl-N′-cyclohexylcarbodiimide, polymer-bound, cross-linked with 2% DVB 100-300 mesh
Synonyms: N-Benzyl-N'-cyclohexylcarbodiimide polymer bound, 1-Benzyl-3-cyclohexylcarbodiimide Resin
00787-25G 00787-1G 00787-5G
220031-061EA 669.23 USD
220031-061 220031-060 220031-062
N-Benzyl-N′-cyclohexylcarbodiimide, polymer-bound, cross-linked with 2% DVB 100-300 mesh
N-Cyclohexylcarbodiimidomethyl polystyrene resin
N-Benzyl-N′-cyclohexylcarbodiimide, polymer-bound
extent of labeling ~1.3 mmol/g N loading
matrix cross-linked with 2% DVB
particle size 100-300 mesh