4-(Trifluoromethyl)phenyl isocyanate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: α,α,α-Trifluoro-p-tolyl isocyanate, 1-Isocyanato-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)phenylisocyanate, p-Trifluoromethylphenylisocyanate

09-7731-1G 09-7731-5G
100197-358EA 63.69 USD
100197-358 100202-362
4-(Trifluoromethyl)phenyl isocyanate ≥99%
4-(Trifluoromethyl)phenyl isocyanate

Organo Fluorine

Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Boiling Pt: 58…59 °C (10 mmHg)
Melting Pt: 58…59 °C
Density: 1.31 g/cm³ (20 °C)
Flash Pt: 158 °F
Storage Temperature: Refrigerator
MDL Number: MFCD00002028
CAS Number: 1548-13-6
EINECS: 216-284-6
UN: 2285
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR