2,2,2-Trifluorothanesulfonyl chloride ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: 2,2,2-Trifluoroethanesulphonyl chloride, (2,2,2-Trifluoroethyl)sulphonyl chloride

09-7304-1G 09-7304-5G 09-7304-250MG
100197-058EA 304.31 USD
100197-058 100201-422 100203-940
2,2,2-Trifluorothanesulfonyl chloride ≥97%
2,2,2-Trifluorothanesulphonyl chloride

Organo Fluorine

Formula: C₂H₂ClF₃O₂S
MW: 182.55 g/mol
Boiling Pt: 140…141 °C
Density: 1.6905 g/cm³ (23 °C)
Flash Pt: 162 °F
MDL Number: MFCD00007458
CAS Number: 1648-99-3
EINECS: 216-713-7
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR