1,1,1-Trifluoroacetylacetone ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
1,1,1-Trifluoropentane-2,4-dione, 1,1,1-Trifluoro-2,4-pentanedione
09-7210-50G
09-7210-10G
100199-222EA
335.45
USD
100199-222
100208-786
1,1,1-Trifluoroacetylacetone ≥98%
1,1,1-Trifluoroacetylacetone
Organo Fluorine
Formula:
C₅H₅F₃O₂ MW: 154.09 g/mol Boiling Pt: 107 °C Density: 1.28 g/cm³ (20 °C) Flash Pt: 79 °F Storage Temperature: Refrigerator |
MDL Number:
MFCD00000427 CAS Number: 367-57-7 EINECS: 206-698-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...