1-(2-Thenoyl)-3,3,3-trifluoroacetone ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

09-6950-100G 09-6950-25G
100208-784EA 409.05 USD
100208-784 100196-018
1-(2-Thenoyl)-3,3,3-trifluoroacetone ≥99%
1-(2-Thenoyl)-3,3,3-trifluoroacetone

Organo Fluorine

Formula: C₈H₅F₃O₂S
Boiling Pt: 96…98 °C (8 mmHg)
Melting Pt: 42…43 °C
Flash Pt: 111 °C
MDL Number: MFCD00005445
CAS Number: 326-91-0
EINECS: 206-316-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR