Tetramethylammonium tetrafluoroborate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tetramethylammoniumtetrafluoroborate
09-6938-10G
09-6938-50G
100204-882EA
83.51
USD
100204-882
100209-630
Tetramethylammonium tetrafluoroborate ≥97%
Tetramethylammonium tetrafluoroborate
Organo Fluorine
Formula:
C₄H₁₂BF₄N Melting Pt: >330 °C |
MDL Number:
MFCD00011745 CAS Number: 661-36-9 EINECS: 211-547-1 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...