2,3,5,6-Tetrafluorothiophenol 98

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1-Mercapto-2,3,5,6-tetrafluorobenzene, 2,3,5,6-Tetrafluorobenzenethiol, 2,3,5,6-Tetrafluorobenzene-1-thiol, 2,3,5,6-tetrafluorothiophenol

09-6920-5G 09-6920-25G
100200-402EA 219.38 USD
100200-402 100208-304
2,3,5,6-Tetrafluorothiophenol 98
2,3,5,6-Tetrafluorothiophenol

Organo Fluorine

Formula: C₆H₂F₄S
MW: 182.14 g/mol
Boiling Pt: 152
Flash Pt: 119 °F
MDL Number: MFCD00004829
CAS Number: 769-40-4
EINECS: 212-210-1
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR