Tetrabutylammonium hexafluorophosphate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

09-6634-25G 09-6634-5G
100199-346EA 106.15 USD
100199-346 100204-878
Tetrabutylammonium hexafluorophosphate ≥98%
Tetrabutylammonium hexafluorophosphate

Organo Fluorine

Formula: C₁₆H₃₆F₆NP
MW: 387.43 g/mol
Melting Pt: 244…246 °C
MDL Number: MFCD00011748
CAS Number: 3109-63-5
EINECS: 221-472-6
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR