Tris(perfluorobutyl)amine ≥85%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

09-6220-25G 09-6220-100G
100209-626EA 121.72 USD
100209-626 100198-568
Tris(perfluorobutyl)amine ≥85%
Tris(perfluorobutyl)amine

Organo Fluorine

Formula: C₁₂F₂₇N
MW: 671.1 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: -55 °C
Density: 1.883 g/cm³ (20 °C)
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR